Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190983.1 | NP_001177912.1 | 4 | 507 | ||
NM_016138.4 | NP_057222.2 | 4 | 558 | |||
GenBank mRNA | AF032900.1 | - | 3 | 374 | ||
AF098948.1 | - | 4 | 516 | |||
AF374413.1 | - | 3 | 447 | |||
AK024291.1 | - | 4 | 539 | |||
AK225115.1 | - | 4 | 507 | |||
AK315470.1 | - | 4 | 558 | |||
AL136647.1 | - | 4 | 510 | |||
BC003185.1 | - | 4 | 549 | 3503763 | ||
BC043272.1 | - | 4 | 1308 | 5297074 |