Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016540.3 | NP_057624.3 | 4 | 2926 | ||
XM_011542559.2 | XP_011540861.1 | 3 | 2860 | |||
GenBank mRNA | AB040973.1 | - | 4 | 2759 |
Set Membership: |
![]() |