Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021020.3 | NP_066300.1 | 3 | 2539 | ||
XM_005273394.3 | XP_005273451.1 | 4 | 2804 | |||
XM_011544383.2 | XP_011542685.1 | 4 | 4742 | |||
XM_011544384.2 | XP_011542686.1 | 4 | 2692 | |||
XM_011544385.2 | XP_011542687.1 | 4 | 2666 | |||
XM_011544386.2 | XP_011542688.1 | 4 | 2891 | |||
GenBank mRNA | AF123659.1 | - | 3 | 2539 |