Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MCF2L SIRNA Assays ›
Gene Symbol:
MCF2L
Gene Name:
MCF.2 cell line derived transforming sequence like
Gene Aliases:
ARHGEF14, DBS, OST
Chromosome Location:
Chr.13: 112894378 - 113099739 on Build GRCh38
UniGene ID:
Hs.170422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137007
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MCF2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112732.2
NP_001106203.2
5
583
NM_001320815.1
NP_001307744.1
5
615
NM_001320816.1
NP_001307745.1
5
792
NM_001320817.1
NP_001307746.1
5
615
NM_024979.4
NP_079255.4
5
615
XM_011537482.1
XP_011535784.1
6
572
XM_011537483.2
XP_011535785.1
6
692
XM_011537484.1
XP_011535786.1
5
1019
XM_011537485.1
XP_011535787.1
5
691
XM_011537486.2
XP_011535788.2
5
707
XM_011537487.1
XP_011535789.1
5
612
XM_011537488.1
XP_011535790.1
5
451
XM_011537490.1
XP_011535792.1
5
985
XM_011537491.1
XP_011535793.1
3
400
XM_017020493.1
XP_016875982.1
5
707
XM_017020494.1
XP_016875983.1
5
707
XM_017020495.1
XP_016875984.1
5
707
XM_017020496.1
XP_016875985.1
3
400
XM_017020497.1
XP_016875986.1
5
985
XM_017020498.1
XP_016875987.1
3
400
XM_017020499.1
XP_016875988.1
5
707
GenBank mRNA
AB002360.1
-
5
560
AB116074.1
-
5
471
AK094768.1
-
3
380
AK295047.1
-
5
612
AK295670.1
-
5
425
AK307867.1
-
5
763
AK315969.1
-
5
583
BC011853.2
-
5
575
4138009
BC020208.1
-
5
575
4121441
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0