Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 10, 11 | 1556 | ||
NM_033624.2 | NP_296373.1 | 10, 11 | 1577 | |||
XM_017019037.1 | XP_016874526.1 | 9, 10 | 1402 | |||
XM_017019038.1 | XP_016874527.1 | 9, 10 | 1381 | |||
XM_017019039.1 | XP_016874528.1 | 9, 10 | 1172 | |||
GenBank mRNA | AB020682.1 | - | 10, 11 | 1518 | ||
AF174601.1 | - | 10, 11 | 1517 | |||
AK001699.1 | - | 10, 11 | 1540 | |||
AK074714.1 | - | 7, 8 | 1025 | |||
AK308141.1 | - | 11, 12 | 1462 | |||
AK314573.1 | - | 10, 11 | 1529 | |||
AL136899.1 | - | 10, 11 | 1390 | |||
BC034045.1 | - | 11, 12 | 1483 | 4815947 | ||
BC042333.1 | - | 10, 11 | 1276 | 5195515 | ||
BC091496.1 | - | 10, 11 | 1515 |