Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175709.3 | NP_783640.1 | 6 | 2374 | ||
XM_005261413.4 | XP_005261470.1 | 6 | 2216 | |||
XM_006724174.3 | XP_006724237.1 | 6 | 2520 | |||
XM_006724175.3 | XP_006724238.1 | 6 | 2519 | |||
XM_006724176.3 | XP_006724239.1 | 6 | 2345 | |||
XM_006724177.3 | XP_006724240.1 | 6 | 2318 | |||
XM_006724178.3 | XP_006724241.1 | 6 | 2243 | |||
XM_011530025.2 | XP_011528327.1 | 6 | 2244 | |||
XM_017028679.1 | XP_016884168.1 | 6 | 2493 | |||
GenBank mRNA | BC051773.1 | - | 6 | 2325 | 6196346 | |
BC128418.1 | - | 1 | 725 |