Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286680.1 | NP_001273609.1 | 7 | 1130 | ||
NM_182795.1 | NP_877724.1 | 6 | 716 | |||
XM_011544360.2 | XP_011542662.1 | 5 | 1352 | |||
XM_011544362.2 | XP_011542664.1 | 7 | 749 | |||
XM_011544363.2 | XP_011542665.1 | 6 | 630 | |||
XM_011544364.2 | XP_011542666.1 | 6 | 1039 | |||
XM_017012947.1 | XP_016868436.1 | 4 | 1242 | |||
XM_017012948.1 | XP_016868437.1 | 7 | 1349 | |||
XM_017012949.1 | XP_016868438.1 | 6 | 1515 | |||
XM_017012950.1 | XP_016868439.1 | 6 | 1223 | |||
GenBank mRNA | AK094267.1 | - | 6 | 1491 | ||
AY262113.1 | - | 7 | 806 | |||
AY262114.1 | - | 6 | 716 |