Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137094
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
142, 143
26083
NM_182961.3
NP_892006.3
142, 143
26392
XM_006715407.1
XP_006715470.1
144, 145
26416
XM_006715408.2
XP_006715471.1
144, 145
26408
XM_006715409.1
XP_006715472.1
143, 144
26395
XM_006715410.2
XP_006715473.1
143, 144
26378
XM_006715411.1
XP_006715474.1
143, 144
26365
XM_006715412.2
XP_006715475.1
143, 144
26363
XM_006715413.2
XP_006715476.1
143, 144
26351
XM_006715414.1
XP_006715477.1
142, 143
26344
XM_006715415.2
XP_006715478.1
142, 143
26309
XM_006715416.2
XP_006715479.1
142, 143
26294
XM_006715417.2
XP_006715480.1
143, 144
26279
XM_006715420.2
XP_006715483.1
143, 144
26267
XM_006715421.2
XP_006715484.1
143, 144
26264
XM_006715422.1
XP_006715485.1
143, 144
26257
XM_006715423.2
XP_006715486.1
144, 145
26420
XM_006715424.2
XP_006715487.1
143, 144
26378
XM_006715425.2
XP_006715488.1
142, 143
26309
XM_011535641.2
XP_011533943.1
144, 145
26417
XM_011535642.2
XP_011533944.1
144, 145
26405
XM_011535643.1
XP_011533945.1
143, 144
26251
XM_011535644.1
XP_011533946.1
130, 131
24467
XM_011535645.2
XP_011533947.1
126, 127
23860
XM_017010608.1
XP_016866097.1
145, 146
26535
XM_017010609.1
XP_016866098.1
144, 145
26342
XM_017010610.1
XP_016866099.1
143, 144
26321
XM_017010611.1
XP_016866100.1
143, 144
26124
XM_017010612.1
XP_016866101.1
142, 143
26048
XM_017010613.1
XP_016866102.1
142, 143
26306
XM_017010614.1
XP_016866103.1
143, 144
26264
XM_017010615.1
XP_016866104.1
141, 142
26153
XM_017010616.1
XP_016866105.1
143, 144
26351
XM_017010617.1
XP_016866106.1
142, 143
26306
XM_017010618.1
XP_016866107.1
142, 143
26294
XM_017010619.1
XP_016866108.1
130, 131
24499
GenBank mRNA
AB018339.2
-
30, 31
5029
AF043290.2
-
4, 5
575
AF444779.1
-
17, 18
2902
AF495910.1
-
142, 143
26392
AF535142.1
-
142, 143
26083
AK122691.1
-
22, 23
3672
AK127076.1
-
27, 28
4438
AK304825.1
-
16, 17
2594
AK316101.1
-
15, 16
2609
AL833657.1
-
7, 8
1145
AY061755.1
-
57, 58
9482
AY061756.1
-
15, 16
2541
AY184203.1
-
14, 15
2406
AY184206.1
-
55, 56
9377
BC090927.1
-
15, 16
2654
BX537837.1
-
24, 25
3970
BX647837.1
-
28, 29
4633
JQ754366.1
-
5, 6
935
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline