Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137127
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
8
6328
NM_001291877.1
NP_001278806.1
8
6475
NM_001291878.1
NP_001278807.1
3
836
NM_001291879.1
NP_001278808.1
5
686
NM_006997.3
NP_008928.1
3
836
NM_206860.2
NP_996742.1
3
836
NM_206861.2
NP_996743.1
6
901
NM_206862.3
NP_996744.3
9
6463
XM_005269388.1
XP_005269445.1
10
6287
XM_005269389.4
XP_005269446.1
10
6811
XM_005269390.1
XP_005269447.1
10
6287
XM_005269391.1
XP_005269448.1
9
6227
XM_005269392.1
XP_005269449.1
10
6287
XM_005269393.1
XP_005269450.1
10
6287
XM_005269394.1
XP_005269451.1
9
6161
XM_005269395.1
XP_005269452.1
9
6152
XM_005269396.1
XP_005269453.1
10
6287
XM_005269397.1
XP_005269454.1
8
6026
XM_005269399.1
XP_005269456.1
9
6152
XM_006717548.1
XP_006717611.1
10
6287
XM_006717552.1
XP_006717615.1
10
6287
XM_006717553.1
XP_006717616.1
10
6287
XM_006717554.1
XP_006717617.1
10
6287
XM_011539126.1
XP_011537428.1
10
6284
XM_011539128.1
XP_011537430.1
8
860
XM_011539129.1
XP_011537431.1
7
725
XM_011539130.1
XP_011537432.1
7
725
XM_011539131.1
XP_011537433.1
3
698
XM_011539132.1
XP_011537434.1
3
698
XM_011539133.1
XP_011537435.1
4
639
XM_011539134.1
XP_011537436.1
4
910
XM_017015484.1
XP_016870973.1
10
6287
XM_017015485.1
XP_016870974.1
10
6287
XM_017015486.1
XP_016870975.1
10
6287
XM_017015487.1
XP_016870976.1
10
6287
XM_017015488.1
XP_016870977.1
8
6026
XM_017015489.1
XP_016870978.1
8
6026
XM_017015491.1
XP_016870980.1
3
838
XM_017015492.1
XP_016870981.1
3
838
XM_017015493.1
XP_016870982.1
3
698
XM_017015494.1
XP_016870983.1
3
838
XM_017015495.1
XP_016870984.1
3
698
XM_017015496.1
XP_016870985.1
4
639
XM_017015497.1
XP_016870986.1
4
910
XM_017015498.1
XP_016870987.1
3
775
XM_017015499.1
XP_016870988.1
5
879
GenBank mRNA
AF095791.2
-
3
423
AF176646.1
-
4
824
AF220152.2
-
1
500
AF528098.1
-
6
882
AF528099.1
-
9
6444
AK295631.1
-
3
619
AL833304.1
-
8
5562
BC039311.1
-
3
836
BC141837.1
-
4
488
BC142673.1
-
4
488
BC144599.1
-
8
6443
9053129
BC144600.1
-
8
6296
9053130
BC150627.1
-
8
6296
9056997
DR001720.1
-
3
672
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0