Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037283.1 | NP_001032360.1 | 7 | 1361 | ||
NM_003751.3 | NP_003742.2 | 7 | 1361 | |||
XM_011515599.1 | XP_011513901.1 | 7 | 1660 | |||
XM_011515600.1 | XP_011513902.1 | 7 | 1660 | |||
XM_011515601.1 | XP_011513903.1 | 8 | 1543 | |||
XM_017012752.1 | XP_016868241.1 | 7 | 1660 | |||
XM_017012753.1 | XP_016868242.1 | 7 | 1660 | |||
GenBank mRNA | AB209381.1 | - | 2 | 378 | ||
AK300967.1 | - | 8 | 1103 | |||
AK302058.1 | - | 8 | 887 | |||
BC001173.1 | - | 7 | 1309 | 3501759 | ||
BC009986.1 | - | 6 | 706 | |||
BC110865.1 | - | 7 | 1287 | 6175284 | ||
U62583.1 | - | 7 | 1331 | |||
U78525.1 | - | 7 | 1353 |