Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF4G3 SIRNA Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
UniGene ID:
Hs.467084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137188
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198801.1
NP_001185730.1
27
4409
NM_001198802.1
NP_001185731.1
27
4319
NM_003760.4
NP_003751.2
24
3935
XM_011542356.2
XP_011540658.1
30
5009
XM_011542361.2
XP_011540663.1
29
4898
XM_011542370.1
XP_011540672.1
30
5012
XM_011542371.1
XP_011540673.1
28
4754
XM_011542374.1
XP_011540676.1
27
4721
XM_011542375.2
XP_011540677.1
27
6309
XM_011542381.2
XP_011540683.1
17
2395
XM_017002672.1
XP_016858161.1
31
6547
XM_017002673.1
XP_016858162.1
29
4991
XM_017002674.1
XP_016858163.1
29
4988
XM_017002675.1
XP_016858164.1
29
4979
XM_017002676.1
XP_016858165.1
28
4958
XM_017002677.1
XP_016858166.1
28
4835
XM_017002678.1
XP_016858167.1
29
4901
XM_017002679.1
XP_016858168.1
29
4898
XM_017002680.1
XP_016858169.1
26
3928
XM_017002681.1
XP_016858170.1
29
4895
XM_017002682.1
XP_016858171.1
28
4877
XM_017002683.1
XP_016858172.1
28
4877
XM_017002684.1
XP_016858173.1
29
6403
XM_017002685.1
XP_016858174.1
28
4868
XM_017002686.1
XP_016858175.1
28
4865
XM_017002687.1
XP_016858176.1
28
4865
XM_017002688.1
XP_016858177.1
25
3894
XM_017002689.1
XP_016858178.1
30
5012
XM_017002690.1
XP_016858179.1
28
4862
XM_017002691.1
XP_016858180.1
27
4847
XM_017002692.1
XP_016858181.1
27
4844
XM_017002693.1
XP_016858182.1
26
4715
XM_017002694.1
XP_016858183.1
28
4787
XM_017002695.1
XP_016858184.1
28
4784
XM_017002696.1
XP_016858185.1
27
4766
XM_017002697.1
XP_016858186.1
27
4763
XM_017002698.1
XP_016858187.1
27
4754
XM_017002699.1
XP_016858188.1
29
4901
XM_017002700.1
XP_016858189.1
26
4733
XM_017002701.1
XP_016858190.1
26
4730
XM_017002702.1
XP_016858191.1
26
4664
XM_017002703.1
XP_016858192.1
24
3825
XM_017002704.1
XP_016858193.1
28
4877
XM_017002705.1
XP_016858194.1
29
4172
XM_017002706.1
XP_016858195.1
28
4061
XM_017002707.1
XP_016858196.1
26
4004
XM_017002708.1
XP_016858197.1
25
3893
XM_017002709.1
XP_016858198.1
27
4756
GenBank mRNA
AB209119.1
-
27
4278
AF012072.2
-
24
3935
AK294883.1
-
18
3182
AK302873.1
-
15
2176
BC094683.1
-
22
2942
6188273
BC136643.1
-
24
3858
9020633
Z34918.1
-
8
1021
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0