Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AP1G2 SIRNA Assays ›
Gene Symbol:
AP1G2
Gene Name:
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases:
G2AD
Chromosome Location:
Chr.14: 23559563 - 23569056 on Build GRCh38
UniGene ID:
Hs.343244
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137370
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AP1G2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282474.1
NP_001269403.1
10, 11
1451
NM_001282475.1
NP_001269404.1
10, 11
1239
NM_003917.4
NP_003908.1
12, 13
1343
XM_005268167.3
XP_005268224.1
11, 12
1466
XM_005268168.4
XP_005268225.1
11, 12
1508
XM_005268169.3
XP_005268226.1
11, 12
1465
XM_005268170.3
XP_005268227.1
11, 12
1465
XM_005268172.3
XP_005268229.1
12, 13
1294
XM_005268173.3
XP_005268230.1
12, 13
1294
XM_005268174.3
XP_005268231.1
10, 11
1341
XM_005268175.3
XP_005268232.1
10, 11
1355
XM_005268177.3
XP_005268234.1
11, 12
1321
XM_005268178.3
XP_005268235.1
11, 12
1212
XM_005268179.3
XP_005268236.1
11, 12
1317
XM_005268180.4
XP_005268237.1
10, 11
1559
XM_005268182.3
XP_005268239.1
11, 12
1188
XM_006720301.3
XP_006720364.1
11, 12
1563
XM_011537283.2
XP_011535585.1
10, 11
1412
XM_011537286.2
XP_011535588.1
3, 4
587
XM_017021740.1
XP_016877229.1
10, 11
1412
XM_017021741.1
XP_016877230.1
8, 9
1316
XM_017021742.1
XP_016877231.1
10, 11
1383
XM_017021743.1
XP_016877232.1
10, 11
1412
XM_017021744.1
XP_016877233.1
12, 13
1295
XM_017021745.1
XP_016877234.1
12, 13
1295
XM_017021746.1
XP_016877235.1
11, 12
1250
XM_017021747.1
XP_016877236.1
11, 12
2642
XM_017021748.1
XP_016877237.1
9, 10
1270
GenBank mRNA
AB015318.1
-
12, 13
1285
AK096034.1
-
1, 2
242
AK226158.1
-
2, 3
273
AK304682.1
-
10, 11
1074
BC051833.1
-
9, 10
1180
5589077
BX161425.1
-
10, 11
1384
BX247963.1
-
8, 9
1299
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
viral process
vesicle-mediated transport
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0