Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGCE SIRNA Assays ›
Gene Symbol:
SGCE
Gene Name:
sarcoglycan epsilon
Gene Aliases:
DYT11, ESG
Chromosome Location:
Chr.7: 94585224 - 94656209 on Build GRCh38
UniGene ID:
Hs.371199
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137371
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGCE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099400.1
NP_001092870.1
3
441
NM_001099401.1
NP_001092871.1
3
441
NM_001301139.1
NP_001288068.1
2
318
NM_003919.2
NP_003910.1
3
441
XM_005250675.4
XP_005250732.1
4
543
XM_005250677.4
XP_005250734.1
4
524
XM_011516663.2
XP_011514965.1
4
524
XM_011516664.2
XP_011514966.1
4
524
XM_011516665.2
XP_011514967.1
3
419
XM_011516666.2
XP_011514968.1
3
419
XM_011516667.2
XP_011514969.1
5
556
XM_011516669.2
XP_011514971.1
4
462
XM_017012763.1
XP_016868252.1
4
392
XM_017012764.1
XP_016868253.1
3
419
XM_017012765.1
XP_016868254.1
5
555
XM_017012766.1
XP_016868255.1
4
392
XM_017012767.1
XP_016868256.1
4
475
XM_017012768.1
XP_016868257.1
4
462
XM_017012769.1
XP_016868258.1
4
462
XM_017012770.1
XP_016868259.1
4
521
GenBank mRNA
AB117974.1
-
3
293
AF036364.1
-
3
369
AF516515.1
-
3
330
AJ000534.1
-
3
398
AK295021.1
-
4
506
AK298220.1
-
2
275
AK313438.1
-
3
380
AY359042.1
-
3
347
BC021709.2
-
3
381
4822278
BI552969.1
-
3
446
KU178449.1
-
3
330
KU178450.1
-
3
330
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
muscle system process
cell-matrix adhesion
muscle organ development
membrane organization
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-9xb8z:80/10.239.165.207:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0