Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF4G3 SIRNA Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
UniGene ID:
Hs.467084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13738
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198801.1
NP_001185730.1
13
2392
NM_001198802.1
NP_001185731.1
14
2413
NM_003760.4
NP_003751.2
11
2029
XM_011542356.2
XP_011540658.1
16
2992
XM_011542361.2
XP_011540663.1
16
2992
XM_011542370.1
XP_011540672.1
16
2995
XM_011542371.1
XP_011540673.1
14
2737
XM_011542374.1
XP_011540676.1
13
2704
XM_011542375.2
XP_011540677.1
13
4292
XM_011542381.2
XP_011540683.1
3
378
XM_017002672.1
XP_016858161.1
17
4530
XM_017002673.1
XP_016858162.1
15
2974
XM_017002674.1
XP_016858163.1
15
2971
XM_017002675.1
XP_016858164.1
15
2962
XM_017002676.1
XP_016858165.1
14
2941
XM_017002677.1
XP_016858166.1
14
2818
XM_017002678.1
XP_016858167.1
16
2995
XM_017002679.1
XP_016858168.1
15
2881
XM_017002680.1
XP_016858169.1
12
1911
XM_017002681.1
XP_016858170.1
15
2878
XM_017002682.1
XP_016858171.1
15
2971
XM_017002683.1
XP_016858172.1
14
2860
XM_017002684.1
XP_016858173.1
15
4386
XM_017002685.1
XP_016858174.1
15
2962
XM_017002686.1
XP_016858175.1
15
2959
XM_017002687.1
XP_016858176.1
14
2848
XM_017002688.1
XP_016858177.1
11
1877
XM_017002689.1
XP_016858178.1
16
2995
XM_017002690.1
XP_016858179.1
14
2845
XM_017002691.1
XP_016858180.1
14
2941
XM_017002692.1
XP_016858181.1
13
2827
XM_017002693.1
XP_016858182.1
12
2698
XM_017002694.1
XP_016858183.1
15
2881
XM_017002695.1
XP_016858184.1
15
2878
XM_017002696.1
XP_016858185.1
14
2860
XM_017002697.1
XP_016858186.1
14
2857
XM_017002698.1
XP_016858187.1
14
2848
XM_017002699.1
XP_016858188.1
16
2995
XM_017002700.1
XP_016858189.1
13
2827
XM_017002701.1
XP_016858190.1
13
2824
XM_017002702.1
XP_016858191.1
12
2647
XM_017002703.1
XP_016858192.1
11
1919
XM_017002704.1
XP_016858193.1
15
2971
XM_017002705.1
XP_016858194.1
15
2155
XM_017002706.1
XP_016858195.1
15
2155
XM_017002707.1
XP_016858196.1
12
1987
XM_017002708.1
XP_016858197.1
12
1987
XM_017002709.1
XP_016858198.1
13
2739
XM_017002710.1
XP_016858199.1
16
2995
XM_017002711.1
XP_016858200.1
16
2995
GenBank mRNA
AB209119.1
-
14
2372
AF012072.2
-
11
2029
AK294883.1
-
5
1276
AK302873.1
-
1
159
BC094683.1
-
9
1036
6188273
BC136643.1
-
11
1952
9020633
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0