Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199096.1 | NP_001186025.1 | 7 | 697 | ||
NM_001199097.1 | NP_001186026.1 | 7 | 639 | |||
NM_001199098.1 | NP_001186027.1 | 7 | 570 | |||
NM_001199099.1 | NP_001186028.1 | 7 | 555 | |||
NM_001286464.1 | NP_001273393.1 | 7 | 690 | |||
NM_003933.4 | NP_003924.2 | 7 | 747 | |||
XM_011522728.1 | XP_011521030.1 | 7 | 1293 | |||
XM_011522729.1 | XP_011521031.1 | 7 | 1293 | |||
XM_011522730.2 | XP_011521032.1 | 7 | 1293 | |||
GenBank mRNA | AB018277.2 | - | 7 | 599 | ||
AK294491.1 | - | 7 | 798 | |||
AK294744.1 | - | 7 | 515 | |||
AK295645.1 | - | 7 | 529 | |||
AK299309.1 | - | 7 | 599 | |||
AK299358.1 | - | 7 | 697 | |||
AK315059.1 | - | 7 | 747 | |||
AL834321.1 | - | 7 | 641 | |||
BC104966.1 | - | 7 | 610 | 8143969 | ||
BC112129.1 | - | 7 | 622 | 8327597 |