Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199096.1 | NP_001186025.1 | 10 | 1007 | ||
NM_001199097.1 | NP_001186026.1 | 10 | 949 | |||
NM_001199098.1 | NP_001186027.1 | 10 | 880 | |||
NM_001199099.1 | NP_001186028.1 | 10 | 865 | |||
NM_001286464.1 | NP_001273393.1 | 10 | 1000 | |||
NM_003933.4 | NP_003924.2 | 10 | 1057 | |||
XM_011522728.1 | XP_011521030.1 | 10 | 1603 | |||
XM_011522729.1 | XP_011521031.1 | 10 | 1603 | |||
XM_011522730.2 | XP_011521032.1 | 10 | 1603 | |||
GenBank mRNA | AB018277.2 | - | 10 | 909 | ||
AK294491.1 | - | 10 | 1108 | |||
AK294744.1 | - | 10 | 825 | |||
AK295645.1 | - | 10 | 839 | |||
AK299309.1 | - | 10 | 909 | |||
AK299358.1 | - | 10 | 1007 | |||
AK315059.1 | - | 10 | 1057 | |||
AL834321.1 | - | 10 | 951 | |||
BC104966.1 | - | 10 | 920 | 8143969 | ||
BC112129.1 | - | 10 | 932 | 8327597 |