Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258288.1 | NP_001245217.1 | 5 | 531 | ||
NM_001258289.1 | NP_001245218.1 | 6 | 926 | |||
NM_003944.3 | NP_003935.2 | 6 | 717 | |||
GenBank mRNA | AK023875.1 | - | 4 | 1099 | ||
AK296661.1 | - | 6 | 895 | |||
AK298355.1 | - | 6 | 582 | |||
AK303815.1 | - | 5 | 497 | |||
AK315643.1 | - | 6 | 687 | |||
BC009084.1 | - | 6 | 671 | 3853674 | ||
BC032997.1 | - | 4 | 1102 | 4839079 | ||
CR456852.1 | - | 5 | 624 | |||
EU794660.1 | - | 6 | 671 | |||
U29091.1 | - | 5 | 628 |