Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003972.2 | NP_003963.1 | 8 | 985 | ||
XM_011540326.1 | XP_011538628.1 | 8 | 1195 | |||
XM_011540327.2 | XP_011538629.1 | 5 | 750 | |||
XM_011540328.2 | XP_011538630.1 | 5 | 578 | |||
XM_011540329.2 | XP_011538631.1 | 8 | 1195 | |||
XM_017016877.1 | XP_016872366.1 | 7 | 1047 | |||
XM_017016878.1 | XP_016872367.1 | 7 | 877 | |||
XM_017016879.1 | XP_016872368.1 | 7 | 873 | |||
XM_017016880.1 | XP_016872369.1 | 7 | 877 | |||
XM_017016881.1 | XP_016872370.1 | 6 | 726 | |||
GenBank mRNA | AF038362.1 | - | 8 | 1004 | ||
AJ001017.2 | - | 8 | 899 | |||
AK308399.1 | - | 8 | 1195 | |||
BC030273.1 | - | 2 | 104 | |||
BC112201.1 | - | 8 | 969 | 8327669 |