Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317800.1 | NP_001304729.1 | 3 | 766 | ||
NM_003974.3 | NP_003965.2 | 5 | 1136 | |||
NM_201349.2 | NP_958728.1 | 4 | 1048 | |||
XM_005273680.4 | XP_005273737.1 | 4 | 863 | |||
GenBank mRNA | AK226110.1 | - | 3 | 736 | ||
AK292126.1 | - | 5 | 1092 | |||
AK296671.1 | - | 4 | 1030 | |||
BC032623.1 | - | 5 | 1104 | 5242017 |