Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003979.3 | NP_003970.1 | 4 | 2431 | ||
GenBank mRNA | AF095448.1 | - | 4 | 1880 | ||
AK001761.1 | - | 4 | 2034 | |||
AK122672.1 | - | 4 | 2671 | |||
AK172760.1 | - | 3 | 1401 | |||
BC003665.2 | - | 4 | 1876 | 2988011 |
Set Membership: |
![]() |