Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004224.3 | NP_004215.2 | 1 | 259 | ||
GenBank mRNA | BC103696.1 | - | 1 | 169 | 40006713 | |
BC105683.1 | - | 1 | 169 | 40006718 | ||
BC105684.1 | - | 1 | 169 | 40006715 | ||
BP245442.1 | - | 1 | 259 | |||
EU432118.1 | - | 1 | 121 | |||
U52219.1 | - | 1 | 190 |
Set Membership: |
![]() |