Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031689.2 | NP_001026859.1 | 4 | 944 | ||
NM_001321546.1 | NP_001308475.1 | 4 | 947 | |||
XM_011518072.2 | XP_011516374.1 | 4 | 933 | |||
XM_017015319.1 | XP_016870808.1 | 4 | 933 | |||
GenBank mRNA | AF083395.1 | - | 3 | 348 | ||
AF145020.1 | - | 4 | 743 | |||
AJ238243.1 | - | 3 | 376 | |||
AK002143.1 | - | 4 | 707 | |||
AK022761.1 | - | 4 | 622 | |||
AK223544.1 | - | 3 | 379 | |||
AL133608.1 | - | 4 | 937 | |||
BC032551.1 | - | 4 | 626 | 5555038 |