Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271811.1 | NP_001258740.1 | 4 | 1318 | ||
NM_004264.4 | NP_004255.2 | 4 | 1320 | |||
NR_073463.1 | 3 | 1205 | ||||
NR_073464.1 | 4 | 1570 | ||||
GenBank mRNA | AK309064.1 | - | 3 | 1177 | ||
BC008380.1 | - | 4 | 1292 | 4041267 |