Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PMPCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004279.2 | NP_004270.2 | 5 | 559 | ||
XM_005250717.3 | XP_005250774.1 | 5 | 573 | |||
XM_006716181.3 | XP_006716244.1 | 5 | 572 | |||
GenBank mRNA | AF054182.1 | - | 5 | 538 | ||
AK000994.1 | - | 5 | 541 | |||
AK057656.1 | - | 4 | 357 | |||
AK098616.1 | - | 5 | 529 | |||
AK289864.1 | - | 5 | 541 | |||
AK297354.1 | - | 5 | 924 | |||
AK309928.1 | - | 529 | ||||
BC014079.2 | - | 5 | 499 | |||
BP311561.1 | - | 5 | 558 |