Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003785.3 | NP_003776.2 | 6 | 565 | ||
XM_011543998.2 | XP_011542300.1 | 6 | 601 | |||
GenBank mRNA | AF058989.1 | - | 6 | 561 | ||
BC004861.2 | - | 6 | 538 | 3829941 | ||
BU941413.1 | - | 6 | 568 |