Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004685.3 | NP_004676.3 | 6 | 994 | ||
XM_011535306.1 | XP_011533608.1 | 6 | 940 | |||
XM_011535307.1 | XP_011533609.1 | 6 | 940 | |||
XM_017020845.1 | XP_016876334.1 | 6 | 940 | |||
XM_017020846.1 | XP_016876335.1 | 2 | 145 | |||
GenBank mRNA | AF072928.1 | - | 2 | 208 | ||
AF406619.1 | - | 6 | 675 | |||
AK314587.1 | - | 6 | 859 | |||
AL832017.2 | - | 6 | 917 | |||
BC040012.1 | - | 6 | 866 | 5495564 | ||
BU177677.1 | - | 1 | 59 | |||
EF491793.1 | - | 1 | 64 |