Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRSF11 SIRNA Assays ›
Gene Symbol:
SRSF11
Gene Name:
serine and arginine rich splicing factor 11
Gene Aliases:
NET2, SFRS11, dJ677H15.2, p54
Chromosome Location:
Chr.1: 70205682 - 70252020 on Build GRCh38
UniGene ID:
Hs.479693
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137702
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SRSF11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190987.1
NP_001177916.1
3
408
NM_004768.3
NP_004759.1
3
411
XM_005271338.3
XP_005271395.1
3
402
XM_005271339.3
XP_005271396.1
3
404
XM_011542427.2
XP_011540729.1
2
564
XM_011542428.2
XP_011540730.1
2
564
XM_011542431.2
XP_011540733.1
3
391
XM_017002811.1
XP_016858300.1
3
402
XM_017002812.1
XP_016858301.1
3
402
XM_017002813.1
XP_016858302.1
3
404
XM_017002815.1
XP_016858304.1
2
564
XM_017002816.1
XP_016858305.1
3
335
XM_017002817.1
XP_016858306.1
2
564
XM_017002818.1
XP_016858307.1
2
564
XM_017002819.1
XP_016858308.1
2
564
XM_017002820.1
XP_016858309.1
2
564
XM_017002821.1
XP_016858310.1
2
564
XM_017002824.1
XP_016858313.1
3
335
XM_017002825.1
XP_016858314.1
2
564
XM_017002826.1
XP_016858315.1
2
564
XM_017002827.1
XP_016858316.1
2
564
XM_017002828.1
XP_016858317.1
3
335
XM_017002829.1
XP_016858318.1
3
371
XM_017002830.1
XP_016858319.1
3
365
XM_017002831.1
XP_016858320.1
2
564
XM_017002832.1
XP_016858321.1
2
564
XM_017002833.1
XP_016858322.1
2
564
XM_017002834.1
XP_016858323.1
2
1523
GenBank mRNA
AK021742.1
-
1
1499
AK300147.1
-
3
355
AK301670.1
-
2
462
AK310447.1
-
3
366
AK310666.1
-
3
335
AK311184.1
-
2
141
BC002784.2
-
2
422
BC009840.2
-
2
405
BC017359.2
-
2
528
BC022815.1
-
3
408
BC032727.1
-
3
395
BC040436.1
-
3
366
5518580
M74002.1
-
3
370
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA processing
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0