Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137726
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
5
1929
NM_001272020.1
NP_001258949.1
4
1839
NM_004796.5
NP_004787.2
15
3292
NM_138970.4
NP_620426.2
4
1839
NR_073546.1
5
1929
NR_073547.1
18
4963
XM_005268218.2
XP_005268275.1
21
5032
XM_006720322.2
XP_006720385.1
20
5020
XM_006720323.2
XP_006720386.1
19
4996
XM_011537363.1
XP_011535665.1
21
5032
XM_011537364.2
XP_011535666.1
22
4994
XM_011537365.2
XP_011535667.1
21
4820
XM_011537366.1
XP_011535668.1
20
5008
XM_011537367.1
XP_011535669.1
20
4942
XM_011537368.1
XP_011535670.1
21
5032
XM_011537369.1
XP_011535671.1
21
5032
XM_011537370.1
XP_011535672.1
20
5008
XM_011537371.1
XP_011535673.1
21
5032
XM_011537372.1
XP_011535674.1
21
5032
XM_011537373.1
XP_011535675.1
20
4942
XM_011537377.2
XP_011535679.1
17
3324
XM_017021790.1
XP_016877279.1
22
5132
XM_017021791.1
XP_016877280.1
22
4915
XM_017021792.1
XP_016877281.1
20
4815
XM_017021793.1
XP_016877282.1
20
5005
XM_017021794.1
XP_016877283.1
19
4996
XM_017021795.1
XP_016877284.1
19
4791
XM_017021796.1
XP_016877285.1
19
4981
XM_017021797.1
XP_016877286.1
18
4972
XM_017021798.1
XP_016877287.1
18
4969
XM_017021799.1
XP_016877288.1
20
4942
XM_017021800.1
XP_016877289.1
20
4942
XM_017021801.1
XP_016877290.1
19
4915
XM_017021802.1
XP_016877291.1
18
4936
XM_017021803.1
XP_016877292.1
20
5011
XM_017021804.1
XP_016877293.1
20
5005
XM_017021805.1
XP_016877294.1
19
4996
XM_017021806.1
XP_016877295.1
19
4981
XM_017021807.1
XP_016877296.1
20
4942
GenBank mRNA
AB018286.1
-
15
3273
AJ316284.2
-
18
4751
AJ493127.1
-
5
1005
AK056530.1
-
4
1839
AL833739.1
-
4
1134
BC059368.1
-
4
1765
BC068469.1
-
4
1765
BC142649.1
-
5
1362
BC150194.1
-
5
1362
BC152457.1
-
15
3273
8862590
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0