Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004860.3 | NP_004851.2 | 15 | 2152 | ||
GenBank mRNA | AK226082.1 | - | 7 | 963 | ||
AK313836.1 | - | 15 | 1837 | |||
BC020090.1 | - | 15 | 2106 | 4385430 | ||
BC051907.1 | - | 15 | 1983 | 6149976 | ||
BC067272.1 | - | 15 | 2097 | 6204341 | ||
BT009817.1 | - | 15 | 1787 | |||
U31501.1 | - | 15 | 2014 |