Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290229.1 | NP_001277158.1 | 7 | 916 | ||
NM_004866.5 | NP_004857.4 | 8 | 994 | |||
NR_110885.1 | 7 | 834 | ||||
XM_011543727.2 | XP_011542029.1 | 7 | 892 | |||
GenBank mRNA | AF005037.1 | - | 8 | 868 | ||
AF038966.1 | - | 8 | 915 | |||
AK001541.1 | - | 2 | 222 | |||
AK225172.1 | - | 8 | 883 | |||
AK290225.1 | - | 8 | 844 | |||
AK298146.1 | - | 6 | 627 | |||
AK304452.1 | - | 7 | 808 | |||
BC009787.1 | - | 7 | 726 | |||
BC015065.1 | - | 8 | 887 | 4105068 | ||
BC016355.1 | - | 2 | 252 | |||
BC034048.1 | - | 8 | 848 | 4814951 | ||
CR542286.1 | - | 8 | 825 |