Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004869.3 | NP_004860.2 | 11 | 2933 | ||
XM_006722582.2 | XP_006722645.1 | 10 | 2809 | |||
XM_011526268.2 | XP_011524570.1 | 12 | 3000 | |||
GenBank mRNA | AF195514.1 | - | 11 | 2874 |