Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005108.3 | NP_005099.2 | 16 | 1438 | ||
XM_011534325.2 | XP_011532627.1 | 16 | 1419 | |||
XM_011534327.1 | XP_011532629.1 | 16 | 1419 | |||
XM_011534328.2 | XP_011532630.1 | 16 | 1419 | |||
XM_011534329.1 | XP_011532631.1 | 16 | 1419 | |||
XM_011534330.2 | XP_011532632.1 | 16 | 1419 | |||
XM_017007595.1 | XP_016863084.1 | 14 | 1187 | |||
XM_017007596.1 | XP_016863085.1 | 16 | 1464 | |||
XM_017007597.1 | XP_016863086.1 | 8 | 753 | |||
XM_017007598.1 | XP_016863087.1 | 16 | 1419 | |||
XM_017007599.1 | XP_016863088.1 | 15 | 1322 | |||
XM_017007600.1 | XP_016863089.1 | 13 | 1232 | |||
GenBank mRNA | AB015046.1 | - | 16 | 1458 | ||
AK001205.1 | - | 9 | 709 | |||
AK025728.1 | - | 16 | 1390 | |||
AK293325.1 | - | 14 | 1192 | |||
AK314386.1 | - | 16 | 1386 | |||
BC137076.1 | - | 16 | 1370 | 9021073 | ||
BC137080.1 | - | 16 | 1370 | 9021077 |