Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173533.1 | NP_001167004.1 | 9 | 2964 | ||
NM_001173534.1 | NP_001167005.1 | 9 | 2955 | |||
NM_001184781.1 | NP_001171710.1 | 10 | 3078 | |||
NM_005137.2 | NP_005128.1 | 10 | 3087 | |||
NR_033674.1 | 9 | 2972 | ||||
GenBank mRNA | BC040500.1 | - | 10 | 3082 | 5272271 | |
CR936871.1 | - | 11 | 3422 | |||
D79985.1 | - | 10 | 3024 | |||
L77560.1 | - | 10 | 2988 | |||
X83545.1 | - | 10 | 2988 | |||
X84076.1 | - | 10 | 2985 |