Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005458.7 | NP_005449.5 | 2 | 798 | ||
GenBank mRNA | AF056085.1 | - | 2 | 795 | ||
AF069755.1 | - | 2 | 494 | |||
AF074483.1 | - | 2 | 338 | |||
AF095784.1 | - | 2 | 469 | |||
AF099033.1 | - | 2 | 338 | |||
AJ012188.1 | - | 2 | 338 | |||
BC035071.2 | - | 3 | 633 | 5260138 | ||
JQ308173.1 | - | 2 | 338 |
Set Membership: |
![]() |