Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005458.7 | NP_005449.5 | 15, 16 | 2697 | ||
XM_005252316.4 | XP_005252373.1 | 13, 14 | 1730 | |||
XM_017015331.1 | XP_016870820.1 | 14, 15 | 2115 | |||
XM_017015332.1 | XP_016870821.1 | 12, 13 | 1684 | |||
GenBank mRNA | AF056085.1 | - | 15, 16 | 2694 | ||
AF069755.1 | - | 15, 16 | 2393 | |||
AF074483.1 | - | 15, 16 | 2237 | |||
AF095784.1 | - | 15, 16 | 2368 | |||
AF099033.1 | - | 15, 16 | 2237 | |||
AJ012188.1 | - | 15, 16 | 2237 | |||
AK308139.1 | - | 14, 15 | 1870 | |||
BC035071.2 | - | 16, 17 | 2532 | 5260138 | ||
JQ308173.1 | - | 15, 16 | 2237 |
Set Membership: |
![]() |