Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005458.7 | NP_005449.5 | 19 | 5775 | ||
GenBank mRNA | AF056085.1 | - | 19 | 5775 | ||
BC055425.1 | - | 1 | 2323 |
Set Membership: |
![]() |