Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI1 SIRNA Assays ›
Gene Symbol:
ABI1
Gene Name:
abl interactor 1
Gene Aliases:
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
Chromosome Location:
Chr.10: 26746596 - 26861087 on Build GRCh38
UniGene ID:
Hs.508148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137946
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012750.2
NP_001012768.1
5, 6
812
NM_001012751.2
NP_001012769.1
5, 6
812
NM_001012752.2
NP_001012770.1
4, 5
797
NM_001178116.1
NP_001171587.1
6, 7
863
NM_001178119.1
NP_001171590.1
5, 6
812
NM_001178120.1
NP_001171591.1
5, 6
812
NM_001178121.1
NP_001171592.1
4, 5
797
NM_001178122.1
NP_001171593.1
5, 6
812
NM_001178123.1
NP_001171594.1
5, 6
812
NM_001178124.1
NP_001171595.1
4, 5
797
NM_001178125.1
NP_001171596.1
3, 4
620
NM_005470.3
NP_005461.2
5, 6
812
XM_005252332.3
XP_005252389.1
4, 5
612
XM_005252333.3
XP_005252390.1
4, 5
612
XM_006717359.2
XP_006717422.1
5, 6
711
XM_006717360.2
XP_006717423.1
5, 6
711
XM_006717361.2
XP_006717424.1
4, 5
612
XM_011519289.2
XP_011517591.1
5, 6
711
XM_017015458.1
XP_016870947.1
5, 6
711
XM_017015459.1
XP_016870948.1
5, 6
711
GenBank mRNA
AB040151.1
-
5, 6
755
AB209268.1
-
5, 6
663
AF001628.1
-
5, 6
636
AF006516.1
-
5, 6
654
AF260262.1
-
4, 5
696
AF540955.1
-
4, 5
585
AK126803.1
-
6, 7
762
AK291823.1
-
4, 5
658
AK296751.1
-
3, 4
362
AK298646.1
-
3, 4
517
BC013238.1
-
5, 6
660
BC024254.2
-
4, 5
648
3531592
FJ380057.1
-
5, 6
588
FJ380058.1
-
5, 6
588
FJ380059.1
-
4, 5
573
U87166.1
-
5, 6
669
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
somitogenesis
movement of cell or subcellular component
transmembrane receptor protein tyrosine kinase signaling pathway
actin polymerization or depolymerization
negative regulation of cell proliferation
viral process
peptidyl-tyrosine phosphorylation
megakaryocyte development
Fc-gamma receptor signaling pathway involved in phagocytosis
vascular endothelial growth factor receptor signaling pathway
positive regulation of protein tyrosine kinase activity
lamellipodium morphogenesis
cell-cell adhesion
protein binding
cytoskeletal protein binding
protein tyrosine kinase activator activity
protein complex binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline