Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005698.3 | NP_005689.2 | 4, 5 | 627 | ||
NM_052837.2 | NP_443069.1 | 3, 4 | 549 | |||
XM_006711106.3 | XP_006711169.1 | 2, 3 | 744 | |||
XM_016999991.1 | XP_016855480.1 | 4, 5 | 945 | |||
GenBank mRNA | AF005039.1 | - | 4, 5 | 489 | ||
BC000161.2 | - | 4, 5 | 501 | 2901296 | ||
BC005135.2 | - | 4, 5 | 474 | 3510027 | ||
BC010505.1 | - | 3, 4 | 418 | 3452347 | ||
BM790077.1 | - | 3, 4 | 523 | |||
BU193466.1 | - | 3, 4 | 549 | |||
CR541757.1 | - | 4, 5 | 393 |