Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308426.1 | NP_001295355.1 | 6 | 1065 | ||
NM_001308427.1 | NP_001295356.1 | 6 | 874 | |||
NM_001308428.1 | NP_001295357.1 | 6 | 865 | |||
NM_001308430.1 | NP_001295359.1 | 5 | 808 | |||
NM_001308431.1 | NP_001295360.1 | 5 | 1025 | |||
NM_001308433.1 | NP_001295362.1 | 5 | 825 | |||
NM_001308434.1 | NP_001295363.1 | 5 | 968 | |||
NM_001308435.1 | NP_001295364.1 | 4 | 864 | |||
NM_005793.4 | NP_005784.1 | 6 | 848 | |||
XM_017005512.1 | XP_016861001.1 | 3 | 1399 | |||
XM_017005513.1 | XP_016861002.1 | 6 | 790 | |||
XM_017005514.1 | XP_016861003.1 | 7 | 1047 | |||
XM_017005515.1 | XP_016861004.1 | 5 | 803 | |||
XM_017005517.1 | XP_016861006.1 | 3 | 565 | |||
XM_017005518.1 | XP_016861007.1 | 4 | 694 | |||
GenBank mRNA | AF051941.1 | - | 6 | 782 | ||
AK222555.1 | - | 5 | 747 | |||
AK294809.1 | - | 5 | 781 | |||
AK297364.1 | - | 4 | 573 | |||
AK308970.1 | - | 3 | 552 | |||
BC001808.2 | - | 6 | 795 | 2966629 | ||
BC012828.2 | - | 6 | 795 | 2966629 |