Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271751.1 | NP_001258680.1 | 8 | 997 | ||
NM_005795.5 | NP_005786.1 | 9 | 1162 | |||
XM_005246231.3 | XP_005246288.2 | 10 | 1206 | |||
XM_005246232.3 | XP_005246289.2 | 11 | 1247 | |||
XM_005246234.3 | XP_005246291.2 | 9 | 1078 | |||
XM_017003151.1 | XP_016858640.1 | 10 | 1137 | |||
GenBank mRNA | AK291630.1 | - | 9 | 1120 | ||
AK292998.1 | - | 8 | 955 | |||
AY389506.1 | - | 6 | 614 | |||
L76380.1 | - | 9 | 1168 | |||
U17473.1 | - | 9 | 1084 |
Set Membership: |
![]() |