Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271751.1 | NP_001258680.1 | 14 | 2099 | ||
NM_005795.5 | NP_005786.1 | 15 | 2264 | |||
XM_005246231.3 | XP_005246288.2 | 16 | 2308 | |||
XM_005246232.3 | XP_005246289.2 | 17 | 2349 | |||
XM_005246234.3 | XP_005246291.2 | 15 | 2180 | |||
XM_017003151.1 | XP_016858640.1 | 16 | 2239 | |||
GenBank mRNA | CB270158.1 | - | 3 | 657 | ||
DA883991.1 | - | 1 | 120 |
Set Membership: |
![]() |