Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53I11 SIRNA Assays ›
Gene Symbol:
TP53I11
Gene Name:
tumor protein p53 inducible protein 11
Gene Aliases:
PIG11
Chromosome Location:
Chr.11: 44932348 - 44951306 on Build GRCh38
UniGene ID:
Hs.554791
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138165
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53I11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258320.1
NP_001245249.1
6
799
NM_001258321.1
NP_001245250.1
4
723
NM_001258322.1
NP_001245251.1
4
653
NM_001258323.2
NP_001245252.1
4
480
NM_001258324.1
NP_001245253.1
3
607
NM_001318384.1
NP_001305313.1
4
488
NM_001318385.1
NP_001305314.1
4
684
NM_001318386.1
NP_001305315.1
4
718
NM_001318389.1
NP_001305318.1
3
405
NM_001318390.1
NP_001305319.1
3
405
NM_006034.4
NP_006025.2
3
405
NR_134612.1
3
405
XM_005253227.4
XP_005253284.1
4
500
XM_005253229.1
XP_005253286.1
3
405
XM_011520475.2
XP_011518777.1
4
508
XM_011520476.2
XP_011518778.1
4
529
XM_011520477.2
XP_011518779.1
4
552
XM_011520478.2
XP_011518780.1
4
461
XM_017018580.1
XP_016874069.1
4
368
XM_017018581.1
XP_016874070.1
3
202
GenBank mRNA
AK026607.1
-
3
379
AK054815.1
-
4
459
AK056379.1
-
4
416
AK292837.1
-
3
217
AL537510.3
-
3
358
BC045666.1
-
6
751
5298525
BU149365.1
-
354
DA115495.1
-
4
268
DA120301.1
-
4
311
DA410127.1
-
4
341
DA697978.1
-
4
338
DA729260.1
-
3
341
DA770050.1
-
4
529
DA778197.1
-
4
352
DA801070.1
-
3
343
DB166321.1
-
3
341
U79302.1
-
3
590
Back To Top
More Information
Gene Ontology Categories:
Function(s)
response to stress
negative regulation of cell proliferation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline