Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MARCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006770.3 | NP_006761.1 | 1, 2 | 221 | ||
XM_011512082.1 | XP_011510384.1 | 1, 2 | 224 | |||
XM_017005171.1 | XP_016860660.1 | 1, 2 | 224 | |||
GenBank mRNA | AF035819.1 | - | 1, 2 | 89 | ||
AK297030.1 | - | 1, 2 | 68 | |||
AK313139.1 | - | 1, 2 | 170 | |||
AX429246.1 | - | 1, 2 | 89 | |||
BC016004.1 | - | 1, 2 | 225 | 4618447 |