Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK11 SIRNA Assays ›
Gene Symbol:
NEK11
Gene Name:
NIMA related kinase 11
Gene Aliases:
-
Chromosome Location:
Chr.3: 131026850 - 131350465 on Build GRCh38
UniGene ID:
Hs.657336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1382
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146003.1
NP_001139475.1
4
624
NM_001321220.1
NP_001308149.1
4
624
NM_001321221.1
NP_001308150.1
6
823
NM_001321222.1
NP_001308151.1
4
624
NM_001321223.1
NP_001308152.1
4
742
NM_001321224.1
NP_001308153.1
3
458
NM_024800.4
NP_079076.3
5
697
NM_145910.3
NP_665917.1
5
697
XM_011513167.2
XP_011511469.1
5
897
XM_011513173.1
XP_011511475.1
3
373
XM_011513174.1
XP_011511476.1
5
643
XM_011513175.1
XP_011511477.1
4
485
XM_017007205.1
XP_016862694.1
6
1000
XM_017007206.1
XP_016862695.1
6
918
XM_017007207.1
XP_016862696.1
6
1134
XM_017007208.1
XP_016862697.1
5
718
XM_017007209.1
XP_016862698.1
5
927
XM_017007210.1
XP_016862699.1
5
783
XM_017007211.1
XP_016862700.1
5
718
XM_017007212.1
XP_016862701.1
5
696
XM_017007213.1
XP_016862702.1
5
696
XM_017007214.1
XP_016862703.1
4
799
XM_017007215.1
XP_016862704.1
5
696
XM_017007216.1
XP_016862705.1
4
592
XM_017007217.1
XP_016862706.1
5
696
XM_017007218.1
XP_016862707.1
7
901
XM_017007219.1
XP_016862708.1
4
395
XM_017007220.1
XP_016862709.1
5
718
XM_017007221.1
XP_016862710.1
5
718
XM_017007222.1
XP_016862711.1
5
718
XM_017007223.1
XP_016862712.1
4
592
XM_017007224.1
XP_016862713.1
6
762
XM_017007225.1
XP_016862714.1
6
775
XM_017007226.1
XP_016862715.1
6
1039
XM_017007227.1
XP_016862716.1
4
519
XM_017007228.1
XP_016862717.1
2
134
XM_017007229.1
XP_016862718.1
5
696
XM_017007230.1
XP_016862719.1
5
874
XM_017007231.1
XP_016862720.1
4
497
XM_017007232.1
XP_016862721.1
5
702
XM_017007233.1
XP_016862722.1
4
452
GenBank mRNA
AB071996.1
-
5
664
AB071997.1
-
5
664
AK123021.1
-
4
1591
AK293261.1
-
3
373
AK297300.1
-
4
624
AK316352.1
-
4
539
AL833472.1
-
4
595
BC028587.2
-
4
741
4829802
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
histone phosphorylation
G2 DNA damage checkpoint
intra-S DNA damage checkpoint
intracellular signal transduction
mitotic cell cycle phase transition
regulation of mitotic cell cycle phase transition
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline