Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007332.2 | NP_015628.2 | 3, 4 | 627 | ||
XM_011517624.2 | XP_011515926.1 | 4, 5 | 5443 | |||
XM_011517625.2 | XP_011515927.1 | 5, 6 | 5602 | |||
XM_017013946.1 | XP_016869435.1 | 4, 5 | 1905 | |||
GenBank mRNA | AK307942.1 | - | 3, 4 | 642 | ||
Y10601.1 | - | 3, 4 | 626 |
Gene Symbol: LOC392232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033867.2 | 3, 4 | 526 | |||
GenBank mRNA | AK309726.1 | - | 4, 5 | 633 |