Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163278.1 | NP_001156750.1 | 8 | 1623 | ||
NM_001163279.1 | NP_001156751.1 | 8 | 1620 | |||
NM_014253.3 | NP_055068.2 | 8 | 1623 | |||
XM_011531230.2 | XP_011529532.1 | 11 | 2264 | |||
XM_017029208.1 | XP_016884697.1 | 11 | 2264 | |||
XM_017029209.1 | XP_016884698.1 | 12 | 2319 | |||
XM_017029210.1 | XP_016884699.1 | 10 | 2192 | |||
XM_017029211.1 | XP_016884700.1 | 11 | 2270 | |||
XM_017029212.1 | XP_016884701.1 | 11 | 1881 | |||
XM_017029213.1 | XP_016884702.1 | 10 | 2193 | |||
XM_017029214.1 | XP_016884703.1 | 11 | 2264 | |||
XM_017029215.1 | XP_016884704.1 | 11 | 2263 | |||
XM_017029216.1 | XP_016884705.1 | 11 | 2263 | |||
GenBank mRNA | AF100772.1 | - | 8 | 1623 | ||
BC140783.1 | - | 8 | 1620 | 9021651 | ||
BC144543.1 | - | 8 | 1617 | 9053070 |