Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAR1 SIRNA Assays ›
Gene Symbol:
BCAR1
Gene Name:
BCAR1, Cas family scaffolding protein
Gene Aliases:
CAS, CAS1, CASS1, CRKAS, P130Cas
Chromosome Location:
Chr.16: 75229030 - 75268053 on Build GRCh38
UniGene ID:
Hs.479747
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138284
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170714.1
NP_001164185.1
8
2546
NM_001170715.1
NP_001164186.1
7
2251
NM_001170716.1
NP_001164187.1
7
2208
NM_001170717.1
NP_001164188.1
7
2324
NM_001170718.1
NP_001164189.1
7
2295
NM_001170719.1
NP_001164190.1
7
2385
NM_001170720.1
NP_001164191.1
8
1956
NM_001170721.1
NP_001164192.1
6
2483
NM_014567.3
NP_055382.2
7
2270
XM_005256258.4
XP_005256315.1
7
2470
XM_005256259.4
XP_005256316.1
7
5382
XM_005256260.4
XP_005256317.1
7
2391
XM_011523475.2
XP_011521777.1
7
2419
XM_017023897.1
XP_016879386.1
7
2977
XM_017023898.1
XP_016879387.1
6
2887
XM_017023899.1
XP_016879388.1
5
2698
GenBank mRNA
AB040024.1
-
7
2265
AF218451.1
-
7
2248
AJ242987.2
-
7
2245
AK026121.1
-
3
754
AK027608.1
-
7
2295
AK074958.1
-
6
1680
AK124526.1
-
7
3690
AK124815.1
-
7
2254
AK124844.1
-
6
2061
AK125719.1
-
3
1729
AK293808.1
-
8
2546
AK294513.1
-
7
2301
AK295809.1
-
7
2180
AK302617.1
-
8
1956
AK314722.1
-
7
2270
AY545071.1
-
6
2178
BC014402.1
-
3
912
BC062556.1
-
6
2125
6427808
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
actin filament organization
cell adhesion
epidermal growth factor receptor signaling pathway
G-protein coupled receptor signaling pathway
integrin-mediated signaling pathway
cell proliferation
insulin receptor signaling pathway
positive regulation of endothelial cell migration
cell migration
positive regulation of cell migration
cellular response to hepatocyte growth factor stimulus
regulation of apoptotic process
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
hepatocyte growth factor receptor signaling pathway
antigen receptor-mediated signaling pathway
T cell receptor signaling pathway
B cell receptor signaling pathway
cell division
cell chemotaxis
signal transducer activity
protein binding
SH3 domain binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline