Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222615.1 | - | 4 | 302 | ||
AK290974.1 | - | 4 | 288 | |||
BC020753.1 | - | 4 | 300 | 4809693 | ||
BC021667.1 | - | 4 | 317 | 3682913 | ||
CR457402.1 | - | 4 | 278 | |||
D14659.1 | - | 4 | 284 |