Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R2 SIRNA Assays ›
Gene Symbol:
PPP6R2
Gene Name:
protein phosphatase 6 regulatory subunit 2
Gene Aliases:
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location:
Chr.22: 50343317 - 50445090 on Build GRCh38
UniGene ID:
Hs.449098
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138323
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242898.1
NP_001229827.1
4, 5
796
NM_001242899.1
NP_001229828.1
4, 5
796
NM_001242900.1
NP_001229829.1
4, 5
796
NM_014678.4
NP_055493.2
4, 5
796
XM_006724431.2
XP_006724494.1
5, 6
907
XM_006724434.1
XP_006724497.1
6, 7
1056
XM_006724435.2
XP_006724498.1
4, 5
781
XM_011530720.2
XP_011529022.1
3, 4
481
XM_011530721.2
XP_011529023.1
3, 4
481
XM_011530722.2
XP_011529024.1
3, 4
481
XM_011530723.2
XP_011529025.1
3, 4
481
XM_011530724.2
XP_011529026.1
3, 4
481
XM_011530726.2
XP_011529028.1
3, 4
481
XM_011530727.2
XP_011529029.1
3, 4
481
XM_011530728.2
XP_011529030.1
3, 4
481
XM_011530729.2
XP_011529031.1
3, 4
481
XM_011530730.2
XP_011529032.1
3, 4
481
XM_011530731.1
XP_011529033.1
3, 4
481
XM_011530732.2
XP_011529034.1
6, 7
957
XM_011530734.2
XP_011529036.1
5, 6
831
XM_011530735.2
XP_011529037.1
4, 5
781
XM_011530736.2
XP_011529038.1
5, 6
907
XM_011530737.2
XP_011529039.1
6, 7
1056
XM_011530738.2
XP_011529040.1
5, 6
859
XM_011530739.2
XP_011529041.1
3, 4
521
XM_011530740.2
XP_011529042.1
5, 6
929
XM_017029116.1
XP_016884605.1
3, 4
481
XM_017029117.1
XP_016884606.1
7, 8
1106
XM_017029118.1
XP_016884607.1
7, 8
1058
XM_017029119.1
XP_016884608.1
6, 7
980
XM_017029120.1
XP_016884609.1
6, 7
957
XM_017029121.1
XP_016884610.1
4, 5
781
XM_017029122.1
XP_016884611.1
5, 6
859
XM_017029123.1
XP_016884612.1
4, 5
781
XM_017029124.1
XP_016884613.1
5, 6
907
XM_017029125.1
XP_016884614.1
4, 5
781
XM_017029126.1
XP_016884615.1
4, 5
781
XM_017029127.1
XP_016884616.1
5, 6
859
XM_017029128.1
XP_016884617.1
5, 6
907
XM_017029129.1
XP_016884618.1
4, 5
781
XM_017029130.1
XP_016884619.1
4, 5
781
XM_017029131.1
XP_016884620.1
5, 6
907
XM_017029132.1
XP_016884621.1
5, 6
859
XM_017029133.1
XP_016884622.1
4, 5
735
XM_017029134.1
XP_016884623.1
4, 5
735
XM_017029135.1
XP_016884624.1
5, 6
785
XM_017029136.1
XP_016884625.1
4, 5
735
XM_017029137.1
XP_016884626.1
4, 5
735
XM_017029138.1
XP_016884627.1
4, 5
735
GenBank mRNA
AB014585.1
-
4, 5
768
AK302472.1
-
4, 5
707
BC000976.4
-
5, 6
785
2900437
BC006568.2
-
4, 5
782
3532211
BC032664.1
-
4, 5
793
5553142
BC041698.1
-
4, 5
763
5732101
BC052995.2
-
4, 5
702
6162781
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-xm8cg:80/10.239.167.213:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0