Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010989.2 | NP_001010989.1 | 6 | 908 | ||
NM_014685.3 | NP_055500.1 | 6 | 911 | |||
GenBank mRNA | AB034989.1 | - | 6 | 854 | ||
AF055001.1 | - | 6 | 851 | |||
AF055003.1 | - | 6 | 651 | |||
AF217990.1 | - | 3 | 1192 | |||
AK223233.1 | - | 6 | 854 | |||
AK291565.1 | - | 6 | 854 | |||
AK292088.1 | - | 6 | 854 | |||
AK302167.1 | - | 5 | 815 | |||
BC000086.1 | - | 6 | 846 | 3504515 | ||
BC008320.1 | - | 6 | 845 | 3350209 | ||
BC032673.1 | - | 6 | 853 | 5575914 | ||
CR457116.1 | - | 6 | 756 | |||
D14695.1 | - | 6 | 849 |